Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lrpap1
Entrez Gene ID
333939
Gene Name
low density lipoprotein receptor-related protein associated protein 1
Gene Aliases
wu:fi20f07, wu:fi22c06
Chromosome Location
Chr.1: 295631 - 305563 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201306.1 NP_958463.1 6 - 7 913 111
GenBank mRNA BC049517.1 - 6 - 7 913 111

View Full Product Details