Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pnp5b
Entrez Gene ID
447889
Gene Name
purine nucleoside phosphorylase 5b
Gene Aliases
zgc:101740
Chromosome Location
Chr.2: 58184937 - 58201177 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004628.1 NP_001004628.1 2 - 3 242 68
GenBank mRNA BC081397.1 - 2 - 3 242 68
BC165322.1 - 2 - 3 226 68

View Full Product Details