Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
illr3
Entrez Gene ID
677740
Gene Name
immune-related, lectin-like receptor 3
Gene Aliases
-
Chromosome Location
Chr.19: 3266957 - 3272998 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044742.1 NP_001038207.1 1 - 2 120 130
XM_021468134.1 XP_021323809.1 1 - 2 174 130
GenBank mRNA AY986757.1 - 1 - 2 120 130
BC163143.1 - 1 - 2 120 130
BC163144.1 - 1 - 2 120 130

View Full Product Details