Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tspan3b
Entrez Gene ID
569864
Gene Name
tetraspanin 3b
Gene Aliases
zgc:162904
Chromosome Location
Chr.7: 45576 - 50894 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082866.1 NP_001076335.1 1 - 2 181 151
GenBank mRNA BC134140.1 - 1 - 2 181 151

View Full Product Details