Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tspan3b
Entrez Gene ID
569864
Gene Name
tetraspanin 3b
Gene Aliases
zgc:162904
Chromosome Location
Chr.7: 45576 - 50894 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082866.1 NP_001076335.1 5 - 6 699 131
GenBank mRNA BC134140.1 - 5 - 6 699 131

View Full Product Details