Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ctsbb
Entrez Gene ID
569298
Gene Name
cathepsin Bb
Gene Aliases
-
Chromosome Location
Chr.20: 52886927 - 52902738 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110478.1 NP_001103948.1 7 - 8 685 66
XM_005160932.4 XP_005160989.1 7 - 8 730 66
GenBank mRNA BC115254.1 - 7 - 8 685 66

View Full Product Details