Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
appa
Entrez Gene ID
58083
Gene Name
amyloid beta (A4) precursor protein a
Gene Aliases
app, im:7044540, wu:fj08a12, wu:fj34d10, wu:fk65e12, zgc:85740
Chromosome Location
Chr.1: 617799 - 656793 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131564.3 NP_571639.1 13 - 14 2091 63
GenBank mRNA AF257742.1 - - 1588 63
AF389401.1 - 13 - 14 1964 63
AJ315636.1 - 8 - 9 1167 63
AJ315637.1 - 6 - 7 823 63
AY271746.1 - 12 - 13 1786 63
BC068375.1 - 13 - 14 1972 63
EH435990.1 - 5 - 6 649 63
JQ994487.1 - 12 - 13 1805 63

View Full Product Details