Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
micall2b
Entrez Gene ID
556298
Gene Name
mical-like 2b
Gene Aliases
MICAL-2, fi19c04, wu:fi19c04, zgc:55983, zgc:66216
Chromosome Location
Chr.1: 9460010 - 9486131 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001312897.1 NP_001299826.1 5 - 6 827 76
NR_132439.1 6 - 7 697 76
GenBank mRNA AL917417.1 - 2 - 3 152 76
BC047798.1 - 6 - 7 697 76
HM151010.1 - 5 - 6 636 76

View Full Product Details