Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
micall2b
Entrez Gene ID
556298
Gene Name
mical-like 2b
Gene Aliases
MICAL-2, fi19c04, wu:fi19c04, zgc:55983, zgc:66216
Chromosome Location
Chr.1: 9460010 - 9486131 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001312897.1 NP_001299826.1 6 - 7 1262 100
NR_132439.1 7 - 8 1132 100
GenBank mRNA BC047798.1 - 7 - 8 1132 100
BC057484.1 - 4 - 5 510 100
BQ616560.1 - 4 - 5 508 100
HM151010.1 - 6 - 7 1071 100

View Full Product Details