Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ubap1
Entrez Gene ID
393183
Gene Name
ubiquitin associated protein 1
Gene Aliases
UBAP-1, zgc:56228
Chromosome Location
Chr.21: 11865957 - 11881908 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328344.1 NP_001315273.1 6 - 7 1407 93
GenBank mRNA BC046008.1 - 7 - 8 869 93

View Full Product Details