Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prmt1
Entrez Gene ID
321974
Gene Name
protein arginine methyltransferase 1
Gene Aliases
fb39h07, hrmt1l2, wu:fb39h07, zf1, zgc:66201
Chromosome Location
Chr.3: 32356833 - 32363021 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200650.2 NP_956944.2 6 - 7 672 92
GenBank mRNA BC044522.1 - 6 - 7 673 92
BC057480.1 - 6 - 7 624 92
EH455208.1 - 6 - 7 624 92
EH509404.1 - 6 - 7 679 92

View Full Product Details