Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trappc6bl
Entrez Gene ID
393634
Gene Name
trafficking protein particle complex 6b-like
Gene Aliases
zgc:66304, zgc:85773
Chromosome Location
Chr.15: 81229 - 88272 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200661.1 NP_956955.1 2 - 3 121 77
GenBank mRNA BC057509.1 - 2 - 3 121 77
BC068383.1 - 2 - 3 163 77

View Full Product Details