Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
fbxo42
Entrez Gene ID
335459
Gene Name
F-box protein 42
Gene Aliases
wu:fj14e08, zgc:64224
Chromosome Location
Chr.22: 191635 - 225386 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212739.1 NP_997904.1 - 355 69
GenBank mRNA BC053311.1 - - 355 69

View Full Product Details