Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnase1
Entrez Gene ID
436947
Gene Name
deoxyribonuclease I
Gene Aliases
zgc:92440
Chromosome Location
Chr.3: 27778976 - 27784386 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002674.1 NP_001002674.1 7 - 8 793 65
GenBank mRNA BC076479.1 - 7 - 8 793 65

View Full Product Details