Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
higd2a
Entrez Gene ID
550334
Gene Name
HIG1 hypoxia inducible domain family, member 2A
Gene Aliases
zgc:112385
Chromosome Location
Chr.14: 51015205 - 51018164 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017641.1 NP_001017641.1 1 - 2 202 104
GenBank mRNA BC093277.1 - 1 - 2 202 104

View Full Product Details