Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
flvcr1
Entrez Gene ID
562595
Gene Name
FLVCR heme transporter 1
Gene Aliases
si:ch211-147d7.6
Chromosome Location
Chr.20: 37843608 - 37862535 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025522.1 NP_001020693.1 4 - 5 1001 91
XM_005160739.3 XP_005160796.1 4 - 5 1428 91
XM_009294763.3 XP_009293038.1 5 - 6 1309 91

View Full Product Details