Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ftr11
Entrez Gene ID
569723
Gene Name
finTRIM family, member 11
Gene Aliases
si:ch211-220f13.2
Chromosome Location
Chr.2: 204232 - 209886 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003197923.4 XP_003197971.1 5 - 6 1574 86
GenBank mRNA BC154553.1 - 5 - 6 1139 86

View Full Product Details