Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tomm34
Entrez Gene ID
323361
Gene Name
translocase of outer mitochondrial membrane 34
Gene Aliases
wu:fb96b08, zC239J9.1, zgc:56645
Chromosome Location
Chr.6: 52226682 - 52234912 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199638.2 NP_955932.2 2 - 3 624 115
GenBank mRNA BC049337.1 - 2 - 3 341 115
EH519797.1 - 4 - 5 546 115

View Full Product Details