Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tomm34
Entrez Gene ID
323361
Gene Name
translocase of outer mitochondrial membrane 34
Gene Aliases
wu:fb96b08, zC239J9.1, zgc:56645
Chromosome Location
Chr.6: 52226682 - 52234912 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199638.2 NP_955932.2 6 - 7 1176 143
GenBank mRNA BC049337.1 - 6 - 7 893 143

View Full Product Details