Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc25a32a
Entrez Gene ID
393226
Gene Name
solute carrier family 25 member 32a
Gene Aliases
fi40c12, mftc, wu:fi40c12, zgc:55610
Chromosome Location
Chr.19: 12937069 - 12965589 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200256.1 NP_956550.1 1 - 2 402 101
XM_009294099.3 XP_009292374.1 1 - 2 971 101
GenBank mRNA BC048057.1 - 1 - 2 402 101
BC164127.1 - 1 - 2 230 101

View Full Product Details