Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
smarce1
Entrez Gene ID
322248
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1
Gene Aliases
baf57, fb33d02, fb54d11, wu:fb33d02, wu:fb54d11
Chromosome Location
Chr.3: 34985856 - 34994988 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201298.1 NP_958455.2 6 - 7 429 91
XM_005163989.4 XP_005164046.1 6 - 7 525 91
XM_005163990.4 XP_005164047.1 6 - 7 513 91
XM_009299517.3 XP_009297792.1 6 - 7 519 91
XM_021469404.1 XP_021325079.1 5 - 6 420 91
XM_021469407.1 XP_021325082.1 5 - 6 408 91
GenBank mRNA BC044363.1 - 6 - 7 429 91
BC066435.1 - 5 - 6 405 91
BC164725.1 - 4 - 5 393 91

View Full Product Details