Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
msrb2
Entrez Gene ID
405857
Gene Name
methionine sulfoxide reductase B2
Gene Aliases
zgc:85965
Chromosome Location
Chr.24: 17040676 - 17047986 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212921.2 NP_998086.2 2 - 3 474 97
GenBank mRNA BC067722.1 - - 316 97
EH490030.1 - 2 - 3 510 97
EH509367.1 - 2 - 3 331 97

View Full Product Details