Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gmfb
Entrez Gene ID
406384
Gene Name
glia maturation factor, beta
Gene Aliases
fc25b01, wu:fa97c03, wu:fc25b01, zgc:56451, zgc:86831
Chromosome Location
Chr.13: 36585547 - 36593618 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213110.1 NP_998275.1 4 - 5 302 144
GenBank mRNA BC049318.1 - 4 - 5 302 144

View Full Product Details