Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
crygmxl2
Entrez Gene ID
554132
Gene Name
crystallin, gamma MX, like 2
Gene Aliases
im:7140756, zgc:110021
Chromosome Location
Chr.12: 30842677 - 30846055 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024427.2 NP_001019598.1 3 - 3 403 154
GenBank mRNA BC095135.1 - 3 - 3 348 154
CN501051.1 - 2 - 2 344 154
CN506249.1 - 3 - 3 420 154

View Full Product Details