Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fstb
Entrez Gene ID
566538
Gene Name
follistatin b
Gene Aliases
fst2, fstl, hm:zeh1606, zeh1606, zgc:171342
Chromosome Location
Chr.10: 35059 - 43676 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109831.1 NP_001103301.1 5 - 5 2049 97
GenBank mRNA BC151865.1 - 5 - 5 2049 97

View Full Product Details