Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
efna3b
Entrez Gene ID
117506
Gene Name
ephrin-A3b
Gene Aliases
D160, efna3, ephrin-A3, zgc:100772
Chromosome Location
Chr.16: 23087390 - 23189869 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131854.3 NP_571929.1 1 - 1 246 71
GenBank mRNA AB051678.1 - 1 - 1 310 71
BC080233.1 - 1 - 1 250 71

View Full Product Details