Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
impa1
Entrez Gene ID
437018
Gene Name
inositol monophosphatase 1
Gene Aliases
zgc:100926
Chromosome Location
Chr.2: 26682927 - 26688922 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002745.1 NP_001002745.1 2 - 3 318 66
XM_005163396.4 XP_005163453.1 2 - 3 335 66
XM_021479900.1 XP_021335575.1 2 - 3 331 66
GenBank mRNA BC076438.1 - 2 - 3 318 66

View Full Product Details