Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gins4
Entrez Gene ID
572562
Gene Name
GINS complex subunit 4 (Sld5 homolog)
Gene Aliases
im:7137690, zgc:100817
Chromosome Location
Chr.8: 52415553 - 52431522 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003546.2 NP_001003546.2 3 - 4 434 93
XM_021478305.1 XP_021333980.1 3 - 4 495 93
GenBank mRNA BC078242.1 - - 476 93
BC131877.1 - - 423 93
EE697537.1 - 3 - 4 453 93

View Full Product Details