Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sgcg
Entrez Gene ID
445292
Gene Name
sarcoglycan, gamma
Gene Aliases
zgc:100924
Chromosome Location
Chr.15: 9327302 - 9347581 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC077085.1 - 8 - 9 1419 59

View Full Product Details