Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pfdn5
Entrez Gene ID
568182
Gene Name
prefoldin 5
Gene Aliases
wu:fa09c11, wu:fb38c05, zgc:100894
Chromosome Location
Chr.6: 235311 - 240343 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111195.1 NP_001104665.1 4 - 5 285 138
GenBank mRNA BC079502.1 - 4 - 5 299 138
BC155162.1 - 4 - 5 285 138

View Full Product Details