Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spata4
Entrez Gene ID
445563
Gene Name
spermatogenesis associated 4
Gene Aliases
zgc:111786
Chromosome Location
Chr.14: 159067 - 163057 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004013.1 NP_001004013.1 5 - 6 759 71
GenBank mRNA AY651920.1 - 5 - 6 759 71
BC093102.1 - 5 - 6 819 71

View Full Product Details