Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fkbp2
Entrez Gene ID
447939
Gene Name
FKBP prolyl isomerase 2
Gene Aliases
zgc:101826
Chromosome Location
Chr.21: 26991196 - 26996037 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004677.2 NP_001004677.2 2 - 3 260 65
GenBank mRNA BC081409.1 - 2 - 3 65
BC155099.1 - 2 - 3 220 65
BC165129.1 - 1 - 2 65
EH538052.1 - 2 - 3 297 65
EH560251.1 - 4 - 5 595 65

View Full Product Details