Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- nitr5
- Entrez Gene ID
-
100216319
- Gene Name
- novel immune-type receptor 5
- Gene Aliases
-
nitr5.1, nitr5.2, nitr5.3, nitr5.4, nitr5.5, nitr5.6, nitr5_1, nitr5_3, nitr5_4, nitr5_5, nitr5a, si:busm1-263j20.5, si:dz105l16.3, si:dz219h16.13, si:dz263j20.5
- Chromosome Location
-
Chr.7: 17568047 - 17571054 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY570233.1 |
- |
3 - 4 |
747 |
140 |
View Full Product Details