Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mettl5
Entrez Gene ID
449776
Gene Name
methyltransferase like 5
Gene Aliases
im:7137598, zgc:103655
Chromosome Location
Chr.9: 48123070 - 48131567 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005949.1 NP_001005949.1 4 - 5 637 57
GenBank mRNA BC083455.1 - 4 - 5 637 57

View Full Product Details