Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snf8
Entrez Gene ID
492771
Gene Name
SNF8 subunit of ESCRT-II
Gene Aliases
zgc:101578
Chromosome Location
Chr.12: 20652963 - 20658285 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007412.1 NP_001007413.1 2 - 3 359 89
GenBank mRNA BC085373.1 - 2 - 3 359 89
BC165511.1 - 2 - 3 138 89

View Full Product Details