Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
napga
Entrez Gene ID
541353
Gene Name
N-ethylmaleimide-sensitive factor attachment protein, gamma a
Gene Aliases
fi35f06, napgl, wu:fi35f06, zgc:110177
Chromosome Location
Chr.24: 945418 - 982488 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013480.2 NP_001013498.2 1 - 2 141 78
GenBank mRNA BC091448.1 - 1 - 2 143 78
BC164747.1 - 1 - 2 84 78

View Full Product Details