Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
napga
Entrez Gene ID
541353
Gene Name
N-ethylmaleimide-sensitive factor attachment protein, gamma a
Gene Aliases
fi35f06, napgl, wu:fi35f06, zgc:110177
Chromosome Location
Chr.24: 945418 - 982488 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013480.2 NP_001013498.2 5 - 6 345 64
XM_017353827.2 XP_017209316.1 5 - 6 299 64
GenBank mRNA BC091448.1 - 5 - 6 347 64
BC164747.1 - 5 - 6 288 64

View Full Product Details