Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ecrg4a
Entrez Gene ID
791762
Gene Name
ECRG4 augurin precursor a
Gene Aliases
zgc:112443
Chromosome Location
Chr.9: 6371919 - 6380653 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017697.1 NP_001017697.1 2 - 3 145 68
GenBank mRNA BC093311.1 - 2 - 3 145 68

View Full Product Details