Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tm4sf5
Entrez Gene ID
436645
Gene Name
transmembrane 4 L six family member 5
Gene Aliases
wu:fd47d06, zgc:92084
Chromosome Location
Chr.7: 21841043 - 21848205 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002372.1 NP_001002372.1 3 - 4 481 130
GenBank mRNA BC075884.1 - 3 - 4 481 130

View Full Product Details