Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nr4a2b
Entrez Gene ID
436679
Gene Name
nuclear receptor subfamily 4, group A, member 2b
Gene Aliases
ngf1b, nr4a2, nurr1, zgc:92696
Chromosome Location
Chr.6: 12462079 - 12465842 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002406.1 NP_001002406.1 4 - 5 1213 63
GenBank mRNA BC076176.1 - 4 - 5 1213 63

View Full Product Details