Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqbp1
Entrez Gene ID
796523
Gene Name
polyglutamine binding protein 1
Gene Aliases
pqbp1l, si:dz150f13.4, zgc:92646
Chromosome Location
Chr.8: 36582728 - 36589390 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002435.1 NP_001002435.1 3 - 4 265 103
GenBank mRNA BC076131.1 - 3 - 4 265 103

View Full Product Details