Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
etnk2
Entrez Gene ID
555330
Gene Name
ethanolamine kinase 2
Gene Aliases
zgc:113386
Chromosome Location
Chr.11: 60725 - 93676 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034175.1 NP_001029347.1 4 - 5 887 134
GenBank mRNA BC096922.1 - 4 - 5 887 134
BC164400.1 - 4 - 5 728 134

View Full Product Details