Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
top1l
Entrez Gene ID
553496
Gene Name
DNA topoisomerase I, like
Gene Aliases
wu:fc66a02, zgc:136349
Chromosome Location
Chr.11: 25296366 - 25311680 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044324.2 NP_001037789.1 19 - 20 2148 68
GenBank mRNA BC117588.1 - 19 - 20 2148 68

View Full Product Details