Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rad51ap1
Entrez Gene ID
555714
Gene Name
RAD51 associated protein 1
Gene Aliases
si:ch211-240j22.5
Chromosome Location
Chr.4: 5196472 - 5203343 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044338.2 NP_001037803.2 5 - 6 630 79
GenBank mRNA BC133972.1 - 5 - 6 630 79

View Full Product Details