Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rad51ap1
- Entrez Gene ID
-
555714
- Gene Name
- RAD51 associated protein 1
- Gene Aliases
-
si:ch211-240j22.5
- Chromosome Location
-
Chr.4: 5196472 - 5203343 on Build GRCz11
- Gene Symbol
- si:ch211-214j24.1
- Entrez Gene ID
-
100034545
- Gene Name
- si:ch211-214j24.1
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BI673060.1 |
- |
3 - 4 |
334 |
130 |
View Full Product Details