Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tm4sf18
Entrez Gene ID
563601
Gene Name
transmembrane 4 L six family member 18
Gene Aliases
si:ch211-284e20.1
Chromosome Location
Chr.22: 38159823 - 38168571 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045022.2 NP_001038487.2 3 - 4 437 98

View Full Product Details