Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc16a4
Entrez Gene ID
567144
Gene Name
solute carrier family 16 member 4
Gene Aliases
zgc:158390
Chromosome Location
Chr.8: 267198 - 300244 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080599.2 NP_001074068.2 6 - 7 1210 69
XM_005167048.4 XP_005167105.1 8 - 9 1717 69
GenBank mRNA BC129219.1 - 6 - 7 1210 69

View Full Product Details