Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
slco1d1
Entrez Gene ID
326845
Gene Name
solute carrier organic anion transporter family, member 1D1
Gene Aliases
wu:fe36b05, zgc:163027
Chromosome Location
Chr.4: 77389667 - 77432607 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348086.1 NP_001335015.1 2 - 3 191 79
XM_021474933.1 XP_021330608.1 3 - 4 209 79
GenBank mRNA BC139695.1 - 2 - 3 218 79
KP294314.1 - 2 - 3 182 79

View Full Product Details