Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lrrc47
Entrez Gene ID
553404
Gene Name
leucine rich repeat containing 47
Gene Aliases
fe50f06, si:ch211-147a11.7, wu:fe50f06
Chromosome Location
Chr.8: 22327269 - 22337594 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110372.1 NP_001103842.1 1 - 2 594 95
GenBank mRNA BC093410.1 - 1 - 2 234 95
BC133856.1 - 1 - 2 513 95
BC154501.1 - 1 - 2 594 95

View Full Product Details