Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
igsf21b
Entrez Gene ID
567714
Gene Name
immunoglobin superfamily, member 21b
Gene Aliases
zgc:162186
Chromosome Location
Chr.23: 21638264 - 21661916 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110473.1 NP_001103943.1 4 - 5 607 81
GenBank mRNA BC134916.1 - 4 - 5 492 81
CU458945.1 - 4 - 5 637 81

View Full Product Details